siRNA Details
Gene Symbol: 1700021K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200038.1 | NP_001186967.1 | 2 | 393 | ||
NM_172615.4 | NP_766203.1 | 2 | 393 | |||
GenBank mRNA | AK030368.1 | - | 2 | 367 | ||
AK042428.1 | - | 2 | 331 | |||
AK122221.1 | - | 2 | 357 | |||
AK170002.1 | - | 2 | 373 | |||
AK170223.1 | - | 2 | 331 | |||
AK170312.1 | - | 2 | 624 | |||
AK171583.1 | - | 2 | 377 | |||
BC057307.1 | - | 2 | 297 | |||
BC060601.1 | - | 2 | 366 | 5706123 | ||
BC067390.1 | - | 2 | 378 | 30432960 |
