siRNA Details
Gene Symbol: Glis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054020.1 | - | 4 | 1282 | ||
AY220846.1 | - | 2 | 327 | |||
BC129906.1 | - | 2 | 305 | |||
BC129907.1 | - | 2 | 305 | |||
DQ871027.1 | - | 4 | 1282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
