siRNA Details
Gene Symbol: Fam178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081225.1 | NP_001074694.1 | 7 | 2256 | ||
GenBank mRNA | AK028324.1 | - | 3 | 206 | ||
AK030951.1 | - | 3 | 572 | |||
AK031688.1 | - | 7 | 2228 | |||
AK032405.1 | - | 3 | 211 | |||
AK036700.1 | - | 5 | 1833 | |||
AK147588.1 | - | 7 | 2729 | |||
AK161809.1 | - | 3 | 619 | |||
BC060679.1 | - | 3 | 173 | |||
BC150999.1 | - | 7 | 2109 | 9087912 | ||
BC157937.1 | - | 7 | 2109 | 9088117 | ||
BC172097.1 | - | 7 | 2109 | 9054383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
