siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 8, 9 | 992 | ||
GenBank mRNA | AK028716.1 | - | 7, 8 | 668 | ||
AK030086.1 | - | 8, 9 | 979 | |||
AK148181.1 | - | 5, 6 | 502 | |||
AK172264.1 | - | 8, 9 | 929 | |||
AK220265.1 | - | 8, 9 | 942 | |||
BC064036.1 | - | 6, 7 | 637 |

More Information
Other Species Targeted

Gene Ontology Categories:
