siRNA Details
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009316.1 | NP_033342.1 | 17 | 2603 | ||
NM_172688.3 | NP_766276.1 | 16 | 2522 | |||
GenBank mRNA | AK046565.1 | - | 16 | 2488 | ||
AK053911.1 | - | 16 | 2508 | |||
AK133642.1 | - | 10 | 1649 | |||
AK137046.1 | - | 16 | 2523 |
