siRNA Details
Gene Symbol: 1700021K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200038.1 | NP_001186967.1 | 20 | 5200 | ||
NM_172615.4 | NP_766203.1 | 19 | 5155 | |||
GenBank mRNA | AK003056.1 | - | 1 | 256 | ||
AK006000.1 | - | 1 | 287 | |||
AK006217.1 | - | 1 | 1220 | |||
AK030368.1 | - | 19 | 5129 | |||
AK122221.1 | - | 20 | 5170 | |||
AK170223.1 | - | 20 | 5051 | |||
BC037474.1 | - | 2 | 2860 | |||
BC057307.1 | - | 20 | 5104 | |||
BC060601.1 | - | 20 | 5173 | 5706123 | ||
BC067390.1 | - | 19 | 5140 | 30432960 |
