siRNA Details
Gene Symbol: Fam178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081225.1 | NP_001074694.1 | 20 | 5537 | ||
GenBank mRNA | AK028324.1 | - | 15 | 3424 | ||
AK030089.1 | - | 7 | 2416 | |||
AK030951.1 | - | 16 | 3853 | |||
AK032405.1 | - | 16 | 3490 | |||
AK147588.1 | - | 20 | 6008 | |||
BC060679.1 | - | 16 | 3454 | |||
BC150999.1 | - | 20 | 5384 | 9087912 | ||
BC157937.1 | - | 20 | 5379 | 9088117 | ||
BC172097.1 | - | 19 | 5317 | 9054383 |

More Information
Other Species Targeted

Gene Ontology Categories:
