siRNA Details
Gene Symbol: ART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130016.2 | NP_001123488.1 | 2, 3 | 211 | ||
NM_001130017.2 | NP_001123489.1 | 2, 3 | 227 | |||
NM_001179.5 | NP_001170.2 | 2, 3 | 211 | |||
XM_005262997.1 | XP_005263054.1 | 1, 2 | 86 | |||
XM_005262999.1 | XP_005263056.1 | 1, 2 | 86 | |||
XM_005263003.1 | XP_005263060.1 | 1, 2 | 86 | |||
XM_005263004.1 | XP_005263061.1 | 1, 2 | 86 | |||
XM_006714220.1 | XP_006714283.1 | 1, 2 | 86 | |||
XM_011531971.1 | XP_011530273.1 | 1, 2 | 86 | |||
XM_017008206.1 | XP_016863695.1 | 1, 2 | 86 | |||
XM_017008207.1 | XP_016863696.1 | 1, 2 | 86 | |||
XM_017008209.1 | XP_016863698.1 | 1, 2 | 86 | |||
GenBank mRNA | AK225860.1 | - | 2, 3 | 207 | ||
AK295305.1 | - | 2, 3 | 193 | |||
AK308810.1 | - | 2, 3 | 193 | |||
AY543657.1 | - | 2, 3 | 203 | |||
AY543658.1 | - | 3, 4 | 237 | |||
AY543659.1 | - | 4, 5 | 318 | |||
AY543660.1 | - | 2, 3 | 196 | |||
BC008397.1 | - | 2, 3 | 198 | 4244549 | ||
BC017913.1 | - | 2, 3 | 198 | 4292261 | ||
BT007249.1 | - | 1, 2 | 77 | |||
CR541664.1 | - | 1, 2 | 77 | |||
DB451386.1 | - | 3, 4 | 257 | |||
U47054.1 | - | 2, 3 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
