siRNA Details
Gene Symbol: Ganc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034155.1 | - | 5 | 1370 | ||
AK036238.1 | - | 5 | 2237 | |||
BC075687.1 | - | 5 | 1507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
