siRNA Details
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172674.2 | NP_766262.2 | 18 | 4947 | ||
GenBank mRNA | AK033017.1 | - | 17 | 4746 | ||
AK035209.1 | - | 1 | 1496 | |||
BC011337.1 | - | 6 | 2908 | |||
BC060121.1 | - | 17 | 4865 | 6413021 | ||
BC086753.1 | - | 1 | 1393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
