siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 9 | 1134 | ||
GenBank mRNA | AK028716.1 | - | 8 | 810 | ||
AK030086.1 | - | 9 | 1121 | |||
AK041411.1 | - | 9 | 1080 | |||
AK148181.1 | - | 6 | 644 | |||
AK172264.1 | - | 9 | 1071 | |||
AK220265.1 | - | 9 | 1084 | |||
BC064036.1 | - | 7 | 779 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008760918, XM_008760917, XM_008760915, XM_008760919, XM_008760914, and others.
Gene Ontology Categories:
