siRNA Details
Gene Symbol: Ddx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172689.3 | NP_766277.3 | 18 | 4348 | ||
GenBank mRNA | AK049305.1 | - | 9 | 2939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
