siRNA Details
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007704.2 | NP_031730.1 | 18 | 2537 | ||
NM_172696.2 | NP_766284.2 | 44 | 5987 | |||
GenBank mRNA | AF060539.1 | - | 18 | 2437 | ||
AK077268.1 | - | 31 | 4269 | |||
AV287690.2 | - | 3 | 598 | |||
BC057124.1 | - | 18 | 2216 | |||
BC082787.1 | - | 33 | 4380 |

More Information
Other Species Targeted

Gene Ontology Categories:
