siRNA Details
Gene Symbol: Efcab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028618.1 | - | 11 | 2370 | ||
AK030011.1 | - | 11 | 2388 | |||
AK080626.1 | - | 11 | 2354 | |||
AK163435.1 | - | 11 | 2366 | |||
AK216349.1 | - | 3 | 337 | |||
BC016074.1 | - | 10 | 1506 | |||
BC028320.1 | - | 8 | 965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
