siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256195.1 | NP_001243124.1 | 19 | 3149 | ||
NM_001256198.1 | NP_001243127.1 | 19 | 3149 | |||
NM_172703.3 | NP_766291.2 | 19 | 3053 | |||
GenBank mRNA | AK029440.1 | - | 2 | 133 | ||
BC023898.1 | - | 6 | 895 | |||
BC024456.1 | - | 7 | 1006 | |||
BC047531.1 | - | 8 | 1220 | |||
BC072600.1 | - | 19 | 3039 | 6410320 | ||
BC075705.1 | - | 4 | 562 | |||
BC079885.1 | - | 19 | 3103 | |||
BC082549.1 | - | 19 | 3113 |
