siRNA Details
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172704.3 | NP_766292.2 | 2 | 148 | ||
GenBank mRNA | AK053156.1 | - | 2 | 130 | ||
AK089983.1 | - | 2 | 140 | |||
AK156793.1 | - | 2 | 130 | |||
AK171781.1 | - | 2 | 129 | |||
AK172542.1 | - | 2 | 129 | |||
BC075619.1 | - | 2 | 124 | |||
BC079852.1 | - | 2 | 118 | |||
BC085257.1 | - | 2 | 122 | 5414963 | ||
BY784439.1 | - | 2 | 158 |
