siRNA Details
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 2, 3 | 381 | ||
GenBank mRNA | AK034358.1 | - | 2, 3 | 413 | ||
AK036245.1 | - | 2, 3 | 457 | |||
AK043026.1 | - | 2, 3 | 445 | |||
AK045969.1 | - | 2, 3 | 415 | |||
AK048738.1 | - | 2, 3 | 401 | |||
AK081041.1 | - | 2, 3 | 426 | |||
AK133610.1 | - | 2, 3 | 382 | |||
AK134808.1 | - | 2, 3 | 442 | |||
AK161902.1 | - | 2, 3 | 442 | |||
AK165601.1 | - | 2, 3 | 435 | |||
BC132290.1 | - | 2, 3 | 357 | 40130567 | ||
BC138012.1 | - | 2, 3 | 357 | 8861031 |
