siRNA Details
Gene Symbol: Sorbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205219.1 | NP_001192148.1 | 9 | 942 | ||
NM_172752.4 | NP_766340.4 | 9 | 942 | |||
GenBank mRNA | AK050951.1 | - | 5 | 627 | ||
AK079130.1 | - | 8 | 838 | |||
AK083429.1 | - | 8 | 860 | |||
AK122369.1 | - | 2 | 110 | |||
AK139388.1 | - | 9 | 933 | |||
AK140498.1 | - | 7 | 807 | |||
AK171632.1 | - | 8 | 776 | |||
BC039163.1 | - | 8 | 714 | |||
BC151038.1 | - | 9 | 812 | 9087951 | ||
BC157944.1 | - | 7 | 557 | 9007279 | ||
BC172140.1 | - | 9 | 812 | 9054810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
