siRNA Details
Gene Symbol: Zfp809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164624.1 | NP_001158096.1 | 3, 4 | 349 | ||
NM_172763.3 | NP_766351.3 | 3, 4 | 358 | |||
GenBank mRNA | AK049344.1 | - | 3, 4 | 310 | ||
AK089532.1 | - | 3, 4 | 513 | |||
BB658893.1 | - | 3, 4 | 513 | |||
BC099418.1 | - | 3, 4 | 290 | 30525533 | ||
BY729060.1 | - | 3, 4 | 359 |
