siRNA Details
Gene Symbol: Dmxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172771.2 | NP_766359.2 | 2 | 297 | ||
GenBank mRNA | AK032099.1 | - | 2 | 243 | ||
AK053672.1 | - | 2 | 298 | |||
AK170274.1 | - | 2 | 284 |

More Information
Other Species Targeted

Gene Ontology Categories:
