siRNA Details
Gene Symbol: Gbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172777.3 | NP_766365.1 | 6 | 904 | ||
GenBank mRNA | AK088861.1 | - | 6 | 890 | ||
AK165231.1 | - | 889 | ||||
BC057170.1 | - | 893 | 4978410 | |||
DQ985742.1 | - | 888 |

More Information
Other Species Targeted

Gene Ontology Categories:
