siRNA Details
Gene Symbol: Gcn1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172719.2 | NP_766307.2 | 40, 41 | 5189 | ||
GenBank mRNA | AB093217.1 | - | 16, 17 | 2382 | ||
AK077013.1 | - | 4, 5 | 511 | |||
AK090137.1 | - | 40, 41 | 5186 | |||
AK147257.1 | - | 40, 41 | 5182 | |||
BC056933.1 | - | 6, 7 | 821 | |||
BC150735.1 | - | 40, 41 | 5192 | 9087646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
