siRNA Details
Gene Symbol: Sorbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205219.1 | NP_001192148.1 | 10 | 996 | ||
NM_172752.4 | NP_766340.4 | 10 | 996 | |||
GenBank mRNA | AK079130.1 | - | 9 | 892 | ||
AK083429.1 | - | 9 | 914 | |||
AK122369.1 | - | 3 | 164 | |||
AK171632.1 | - | 9 | 830 | |||
BC039163.1 | - | 9 | 768 | |||
BC151038.1 | - | 10 | 866 | 9087951 | ||
BC172140.1 | - | 10 | 866 | 9054810 |

More Information
Other Species Targeted

Gene Ontology Categories:
