siRNA Details
Gene Symbol: Zfp809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164624.1 | NP_001158096.1 | 5 | 480 | ||
NM_172763.3 | NP_766351.3 | 5 | 489 | |||
GenBank mRNA | AK044107.1 | - | 3 | 276 | ||
AK049344.1 | - | 5 | 441 | |||
AK089532.1 | - | 5 | 644 | |||
BC099418.1 | - | 5 | 421 | 30525533 | ||
BY729060.1 | - | 5 | 490 | |||
CK389669.1 | - | 2 | 121 |
