siRNA Details
Gene Symbol: Ttc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172770.3 | NP_766358.1 | 5 | 379 | ||
GenBank mRNA | AK054350.1 | - | 5 | 380 | ||
AK076457.1 | - | 4 | 870 | |||
BC056616.1 | - | 4 | 307 | 4974294 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001004246, XM_006243004, XM_008766196, XM_006243005, BC079409.
Gene Ontology Categories:
