siRNA Details
Gene Symbol: Dmxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172771.2 | NP_766359.2 | 5 | 612 | ||
GenBank mRNA | AK032099.1 | - | 5 | 558 | ||
AK048831.1 | - | 3 | 1297 | |||
AK053672.1 | - | 5 | 613 | |||
AK170274.1 | - | 5 | 599 | |||
AK170407.1 | - | 3 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
