siRNA Details
Gene Symbol: 4932414J04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030001.1 | - | 2 | 481 | ||
BC109353.1 | - | 1 | 272 | 40057015 | ||
BC116680.1 | - | 1 | 272 | 40106469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
