siRNA Details
Gene Symbol: Gcn1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172719.2 | NP_766307.2 | 58 | 8519 | ||
GenBank mRNA | AB093217.1 | - | 34 | 5712 | ||
AK077013.1 | - | 22 | 3838 | |||
AK090137.1 | - | 58 | 8516 | |||
AK147257.1 | - | 58 | 8512 | |||
BC038407.1 | - | 11 | 2256 | |||
BC056933.1 | - | 24 | 4151 | |||
BC068244.1 | - | 14 | 2764 | |||
BC082548.1 | - | 35 | 5538 | |||
BY582410.1 | - | 1 | 371 |

More Information
Other Species Targeted

Gene Ontology Categories:
