siRNA Details
Gene Symbol: E130309D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172726.4 | NP_766314.3 | 4 | 918 | ||
GenBank mRNA | AK041667.1 | - | 4 | 864 | ||
AK050733.1 | - | 4 | 832 | |||
AK053801.1 | - | 4 | 919 | |||
AK054218.1 | - | 4 | 619 | |||
AK087531.1 | - | 4 | 919 | |||
AK172614.1 | - | 4 | 865 | |||
AK198635.1 | - | 2 | 105 | |||
BC058940.1 | - | 3 | 2997 | |||
BC079853.1 | - | 4 | 852 | 5716912 | ||
BC082321.1 | - | 4 | 662 | 6411746 |
