siRNA Details
Gene Symbol: Sorbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205219.1 | NP_001192148.1 | 18, 19 | 3339 | ||
NM_172752.4 | NP_766340.4 | 17, 18 | 3285 | |||
GenBank mRNA | AK050951.1 | - | 13, 14 | 1560 | ||
AK083429.1 | - | 15, 16 | 1619 | |||
AK122369.1 | - | 13, 14 | 2750 | |||
AK140498.1 | - | 13, 14 | 1524 | |||
BC031203.1 | - | 1, 2 | 57 | |||
BC039163.1 | - | 17, 18 | 1689 | |||
BC151038.1 | - | 18, 19 | 3209 | 9087951 | ||
BC157944.1 | - | 14, 15 | 1430 | 9007279 | ||
BC172140.1 | - | 17, 18 | 3155 | 9054810 |

More Information
Other Species Targeted

Gene Ontology Categories:
