siRNA Details
Gene Symbol: Gbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039646.2 | NP_001034735.2 | 10, 11 | 1744 | ||
GenBank mRNA | BC112328.1 | - | 10, 11 | 1761 | 5348193 | |
BC114209.1 | - | 10, 11 | 1744 | 4980612 | ||
DQ985743.1 | - | 10, 11 | 1744 |
Gene Symbol: Gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194336.2 | NP_919317.2 | 10, 11 | 1738 | ||
GenBank mRNA | AK128993.1 | - | 1734 | |||
AK129021.1 | - | 1010 | ||||
AK171913.1 | - | 7, 8 | 3376 | |||
BC007143.1 | - | 9, 10 | 1595 | 3495728 | ||
BC052867.1 | - | 5, 6 | 966 | |||
BC057969.1 | - | 1724 | 4978977 | |||
BC115768.1 | - | 1734 | 40085916 |
Gene Symbol: Gbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172777.3 | NP_766365.1 | 10, 11 | 1770 | ||
GenBank mRNA | AK088861.1 | - | 10, 11 | 1756 | ||
AK165231.1 | - | 2417 | ||||
BC057170.1 | - | 1759 | 4978410 | |||
DQ985742.1 | - | 1754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
