siRNA Details
Gene Symbol: Zfp454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172794.2 | NP_766382.2 | 6 | 1924 | ||
GenBank mRNA | AK046117.1 | - | 3 | 1673 | ||
BC049793.1 | - | 6 | 1924 | 6510760 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172794.2 | NP_766382.2 | 6 | 1924 | ||
GenBank mRNA | AK046117.1 | - | 3 | 1673 | ||
BC049793.1 | - | 6 | 1924 | 6510760 |