siRNA Details
Gene Symbol: Lrrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172879.3 | NP_766467.2 | 2 | 503 | ||
GenBank mRNA | AK029981.1 | - | 2 | 504 | ||
BC141026.1 | - | 2 | 330 | 9055638 | ||
BC145170.1 | - | 2 | 330 | 9053721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
