siRNA Details
Gene Symbol: Sdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177879.5 | NP_808547.3 | 36, 37 | 5474 | ||
GenBank mRNA | AK032883.1 | - | 11, 12 | 1374 | ||
AK035225.1 | - | 2, 3 | 230 | |||
AK147578.1 | - | 36, 37 | 5519 | |||
AK148648.1 | - | 2, 3 | 208 | |||
AY353236.1 | - | 32, 33 | 4624 | |||
BC060237.1 | - | 36, 37 | 5058 | |||
BC138453.1 | - | 32, 33 | 4495 | 8861473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
