siRNA Details
Gene Symbol: Fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172887.2 | NP_766475.2 | 2 | 534 | ||
GenBank mRNA | AK086708.1 | - | 2 | 422 | ||
AK157981.1 | - | 2 | 271 |

More Information
Other Species Targeted

Gene Ontology Categories:
