siRNA Details
Gene Symbol: Sdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177879.5 | NP_808547.3 | 36, 37 | 5483 | ||
GenBank mRNA | AK032883.1 | - | 11, 12 | 1383 | ||
AK035225.1 | - | 2, 3 | 239 | |||
AK147578.1 | - | 36, 37 | 5528 | |||
AK148648.1 | - | 2, 3 | 217 | |||
AY353236.1 | - | 32, 33 | 4633 | |||
BC060237.1 | - | 36, 37 | 5067 | |||
BC138453.1 | - | 32, 33 | 4504 | 8861473 |

More Information
Other Species Targeted

Gene Ontology Categories:
