siRNA Details
Gene Symbol: Rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172866.3 | NP_766454.1 | 8 | 1166 | ||
GenBank mRNA | AK089288.1 | - | 8 | 949 | ||
AK129103.1 | - | 8 | 952 | |||
AK170117.1 | - | 8 | 963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
