siRNA Details
Gene Symbol: Dmkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166173.1 | NP_001159645.1 | 8, 9 | 1205 | ||
GenBank mRNA | AK081753.1 | - | 8, 9 | 1207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
