siRNA Details
Gene Symbol: Hydin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172916.2 | NP_766504.3 | 3 | 373 | ||
GenBank mRNA | AK043971.1 | - | 2 | 192 | ||
AK142981.1 | - | 3 | 360 | |||
AY173049.1 | - | 3 | 373 |

More Information
Other Species Targeted

Gene Ontology Categories:
