siRNA Details
Gene Symbol: Rnf213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077880.1 | - | 7 | 1765 | ||
AK150249.1 | - | 2 | 964 | |||
AK156571.1 | - | 8 | 1898 | |||
AK173199.1 | - | 17 | 5111 | |||
AV319021.2 | - | 1 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
