siRNA Details
Gene Symbol: Idh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173011.2 | NP_766599.2 | 2, 3 | 283 | ||
GenBank mRNA | AF212319.1 | - | 2, 3 | 254 | ||
AK088110.1 | - | 2, 3 | 255 | |||
AK145753.1 | - | 2, 3 | 228 | |||
AK146564.1 | - | 2, 3 | 266 | |||
AK161640.1 | - | 2, 3 | 267 | |||
AK169395.1 | - | 2, 3 | 280 | |||
BC060030.1 | - | 2, 3 | 271 | 6515657 | ||
BY311397.1 | - | 2, 3 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
