siRNA Details
Gene Symbol: Myo9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173018.2 | NP_766606.2 | 31 | 6128 | ||
GenBank mRNA | AK028873.1 | - | 2 | 131 | ||
BC068298.1 | - | 5 | 566 |

More Information
Other Species Targeted

Gene Ontology Categories:
