siRNA Details
Gene Symbol: Tstd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173033.3 | NP_766621.3 | 2, 3 | 312 | ||
GenBank mRNA | AK028258.1 | - | 2, 3 | 255 | ||
AK155456.1 | - | 2, 3 | 259 | |||
AK165359.1 | - | 2, 3 | 251 | |||
BY013797.1 | - | 2, 3 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
