siRNA Details
Gene Symbol: Abcc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172912.4 | NP_766500.3 | 4, 5 | 696 | ||
GenBank mRNA | AF514414.1 | - | 4, 5 | 696 | ||
AF514415.1 | - | 4, 5 | 696 | |||
AK133175.1 | - | 786 | ||||
BC138380.1 | - | 4, 5 | 689 | 8861400 | ||
BC138381.1 | - | 4, 5 | 689 | 8861401 | ||
BC171952.1 | - | 4, 5 | 689 | 9054238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
