siRNA Details
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172938.3 | NP_766526.3 | 7 | 1411 | ||
GenBank mRNA | AK034171.1 | - | 7 | 1250 | ||
AK040597.1 | - | 2 | 1059 | |||
AK043500.1 | - | 2 | 942 | |||
AK047972.1 | - | 5 | 837 | |||
AK141826.1 | - | 2 | 435 | |||
AK172160.1 | - | 3 | 474 | |||
AK220309.1 | - | 5 | 795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
