siRNA Details
Gene Symbol: Myo9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173018.2 | NP_766606.2 | 32, 33 | 6401 | ||
GenBank mRNA | AK028873.1 | - | 3, 4 | 404 | ||
AK029836.1 | - | 2, 3 | 268 | |||
BC068298.1 | - | 6, 7 | 839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
