siRNA Details
Gene Symbol: Hydin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172916.2 | NP_766504.3 | 9 | 1291 | ||
GenBank mRNA | AK043971.1 | - | 8 | 1110 | ||
AK142981.1 | - | 9 | 1278 | |||
AY173049.1 | - | 9 | 1291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
