siRNA Details
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172938.3 | NP_766526.3 | 9 | 3176 | ||
GenBank mRNA | AK034171.1 | - | 9 | 3015 | ||
AK047972.1 | - | 7 | 2602 | |||
AK141826.1 | - | 4 | 2201 | |||
AK172160.1 | - | 5 | 2239 | |||
AK220309.1 | - | 7 | 2560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
