siRNA Details
Gene Symbol: Sowahc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172939.3 | NP_766527.3 | 1 | 1711 | ||
GenBank mRNA | AK030944.1 | - | 1 | 1712 | ||
AK050497.1 | - | 1 | 1682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
