siRNA Details
Gene Symbol: Myo9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173018.2 | NP_766606.2 | 42, 43 | 7565 | ||
GenBank mRNA | AK028873.1 | - | 12, 13 | 1514 | ||
AK029836.1 | - | 11, 12 | 1378 | |||
BC046526.1 | - | 5, 6 | 627 | |||
BC068298.1 | - | 15, 16 | 1946 | |||
BC096035.1 | - | 4, 5 | 574 |

More Information
Other Species Targeted

Gene Ontology Categories:
